Anti-APH1 Mouse Monoclonal Antibody [clone: 66544]

Supplier: R & D Systems
MAB6908
102886-744EA 561.11 USD
102886-744
Anti-APH1 Mouse Monoclonal Antibody [clone: 66544]
Antibodies
Type: Primary
Antigen: APH1
Clonality: Monoclonal
Clone: 66544
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR