Anti-CREB1 Mouse Monoclonal Antibody [clone: 702710]

Supplier: R & D Systems
MAB6906
102886-742EA 594.37 USD
102886-742
Anti-CREB1 Mouse Monoclonal Antibody [clone: 702710]
Antibodies
Type: Primary
Antigen: CREB1
Clonality: Monoclonal
Clone: 702710
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR