Anti-TNF Mouse Monoclonal Antibody [clone: 103302]

Supplier: R & D Systems
MAB6903
220021-442EA 594.37 USD
220021-442
Anti-TNF Mouse Monoclonal Antibody [clone: 103302]
Antibodies
Type: Primary
Antigen: TNF
Clonality: Monoclonal
Clone: 103302
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR