Anti-TNF Mouse Monoclonal Antibody [clone: 103304]

Supplier: R & D Systems
MAB6902
220021-441EA 772.25 USD
220021-441
Anti-TNF Mouse Monoclonal Antibody [clone: 103304]
Antibodies
Type: Primary
Antigen: TNF
Clonality: Monoclonal
Clone: 103304
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR