Anti-TNF Mouse Monoclonal Antibody [clone: 103314]

Supplier: R & D Systems
MAB6901
220021-440EA 763.57 USD
220021-440
Anti-TNF Mouse Monoclonal Antibody [clone: 103314]
Antibodies
Type: Primary
Antigen: TNF
Clonality: Monoclonal
Clone: 103314
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR