Anti-TNF Mouse Monoclonal Antibody [clone: 77938]

Supplier: R & D Systems
MAB690
220021-439EA 763.57 USD
220021-439
Anti-TNF Mouse Monoclonal Antibody [clone: 77938]
Antibodies
Type: Primary
Antigen: TNF
Clonality: Monoclonal
Clone: 77938
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR