Anti-ACKR4 Mouse Monoclonal Antibody [clone: 674144]

Supplier: R & D Systems
MAB6899
102886-738EA 577.02 USD
102886-738
Anti-ACKR4 Mouse Monoclonal Antibody [clone: 674144]
Antibodies
Type: Primary
Antigen: ACKR4
Clonality: Monoclonal
Clone: 674144
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR