Anti-ACACA Mouse Monoclonal Antibody [clone: 738421]

Supplier: R & D Systems
MAB6898
102886-736EA 594.37 USD
102886-736
Anti-ACACA Mouse Monoclonal Antibody [clone: 738421]
Antibodies
Type: Primary
Antigen: ACACA
Clonality: Monoclonal
Clone: 738421
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR