Anti-VCL Mouse Monoclonal Antibody [clone: 728513]

Supplier: R & D Systems
MAB68961
102886-734EA 594.37 USD
102886-734
Anti-VCL Mouse Monoclonal Antibody [clone: 728513]
Antibodies
Type: Primary
Antigen: VCL
Clonality: Monoclonal
Clone: 728513
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR