Anti-VCL Mouse Monoclonal Antibody [clone: 728526]

Supplier: R & D Systems
MAB6896
102886-732EA 655.11 USD
102886-732
Anti-VCL Mouse Monoclonal Antibody [clone: 728526]
Antibodies
Simple Western

Type: Primary
Antigen: VCL
Clonality: Monoclonal
Clone: 728526
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR