Anti-GSTM1 Mouse Monoclonal Antibody [clone: 842516]

Supplier: R & D Systems
MAB6894
102886-730EA 594.37 USD
102886-730
Anti-GSTM1 Mouse Monoclonal Antibody [clone: 842516]
Antibodies
Type: Primary
Antigen Symbol: GSTM1
Antigen Name: glutathione S-transferase mu 1
Antigen Synonyms: GST1, GSTM1-1, GSTM1a-1a, GSTM1b-1b, GTH4, GTM1, H-B, MU, MU-1
Clonality: Monoclonal
Clone: 842516
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR