Anti-AKT1S1 Mouse Monoclonal Antibody [clone: 7652]

Supplier: R & D Systems
MAB6890
102886-728EA 594.37 USD
102886-728
Anti-AKT1S1 Mouse Monoclonal Antibody [clone: 7652]
Antibodies
Type: Primary
Antigen: AKT1S1
Clonality: Monoclonal
Clone: 7652
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR