Anti-TNNI3 Mouse Monoclonal Antibody [clone: 679021]

Supplier: R & D Systems
MAB68871
102886-722EA 577.02 USD
102886-722
Anti-TNNI3 Mouse Monoclonal Antibody [clone: 679021]
Antibodies
Simple Western

Type: Primary
Antigen: TNNI3
Clonality: Monoclonal
Clone: 679021
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR