Anti-AAK1 Mouse Monoclonal Antibody [clone: 72425]

Supplier: R & D Systems
MAB6886
102886-718EA 594.37 USD
102886-718
Anti-AAK1 Mouse Monoclonal Antibody [clone: 72425]
Antibodies
Type: Primary
Antigen: AAK1
Clonality: Monoclonal
Clone: 72425
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR