Anti-EIF2AK4 Mouse Monoclonal Antibody [clone: 708210]

Supplier: R & D Systems
MAB6878
102886-716EA 594.37 USD
102886-716
Anti-EIF2AK4 Mouse Monoclonal Antibody [clone: 708210]
Antibodies
Type: Primary
Antigen: EIF2AK4
Clonality: Monoclonal
Clone: 708210
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR