Anti-KLRC1 Rat Monoclonal Antibody [clone: 705829]

Supplier: R & D Systems
MAB6867
102886-714EA 594.37 USD
102886-714
Anti-KLRC1 Rat Monoclonal Antibody [clone: 705829]
Antibodies
Type: Primary
Antigen: KLRC1
Clonality: Monoclonal
Clone: 705829
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR