Anti-OPRM1 Mouse Monoclonal Antibody [clone: 677014]

Supplier: R & D Systems
MAB6866
102886-712EA 584.25 USD
102886-712
Anti-OPRM1 Mouse Monoclonal Antibody [clone: 677014]
Antibodies
Type: Primary
Antigen: OPRM1
Clonality: Monoclonal
Clone: 677014
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR