Anti-IL6 Mouse Monoclonal Antibody [clone: 90613]

Supplier: R & D Systems
MAB6861
220021-436EA 594.37 USD
220021-436
Anti-IL6 Mouse Monoclonal Antibody [clone: 90613]
Antibodies
Type: Primary
Antigen: IL6
Clonality: Monoclonal
Clone: 90613
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR