Anti-NEU1 Mouse Monoclonal Antibody [clone: 688215]

Supplier: R & D Systems
MAB6860
102886-710EA 594.37 USD
102886-710
Anti-NEU1 Mouse Monoclonal Antibody [clone: 688215]
Antibodies
Type: Primary
Antigen Symbol: NEU1
Antigen Name: sialidase 1 (lysosomal sialidase)
Antigen Synonyms: NANH, NEU, SIAL1
Clonality: Monoclonal
Clone: 688215
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR