Anti-IL6 Mouse Monoclonal Antibody [clone: 77830]

Supplier: R & D Systems
MAB686
220021-435EA 763.57 USD
220021-435
Anti-IL6 Mouse Monoclonal Antibody [clone: 77830]
Antibodies
Type: Primary
Antigen: IL6
Clonality: Monoclonal
Clone: 77830
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR