Anti-BORA Mouse Monoclonal Antibody [clone: 694746]

Supplier: R & D Systems
MAB68561
102886-704EA 561.11 USD
102886-704
Anti-BORA Mouse Monoclonal Antibody [clone: 694746]
Antibodies
Type: Primary
Antigen: BORA
Clonality: Monoclonal
Clone: 694746
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR