Anti-FGFR4 Rat Monoclonal Antibody [clone: 240929]

Supplier: R & D Systems
MAB6852
220021-434EA 633.42 USD
220021-434
Anti-FGFR4 Rat Monoclonal Antibody [clone: 240929]
Antibodies
Type: Primary
Antigen Symbol: FGFR4
Antigen Name: fibroblast growth factor receptor 4
Antigen Synonyms: CD334, JTK2, TKF
Clonality: Monoclonal
Clone: 240929
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR