Anti-DCTN2 Mouse Monoclonal Antibody [clone: 696206]

Supplier: R & D Systems
MAB6850
102886-700EA 561.11 USD
102886-700
Anti-DCTN2 Mouse Monoclonal Antibody [clone: 696206]
Antibodies
Type: Primary
Antigen: DCTN2
Clonality: Monoclonal
Clone: 696206
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR