Anti-PAK2 Mouse Monoclonal Antibody [clone: 677411]

Supplier: R & D Systems
MAB6849
102886-698EA 594.37 USD
102886-698
Anti-PAK2 Mouse Monoclonal Antibody [clone: 677411]
Antibodies
Type: Primary
Antigen: PAK2
Clonality: Monoclonal
Clone: 677411
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR