Anti-STX1B Mouse Monoclonal Antibody [clone: 676204]

Supplier: R & D Systems
MAB6848
102886-696EA 594.37 USD
102886-696
Anti-STX1B Mouse Monoclonal Antibody [clone: 676204]
Antibodies
Type: Primary
Antigen: STX1B
Clonality: Monoclonal
Clone: 676204
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR