Anti-SLC32A1 Mouse Monoclonal Antibody [clone: 731307]

Supplier: R & D Systems
MAB6847
102886-694EA 577.02 USD
102886-694
Anti-SLC32A1 Mouse Monoclonal Antibody [clone: 731307]
Antibodies
Type: Primary
Antigen: SLC32A1
Clonality: Monoclonal
Clone: 731307
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR