Anti-CRTC3 Rat Monoclonal Antibody [clone: 643729]

Supplier: R & D Systems
MAB6840
102886-692EA 561.11 USD
102886-692
Anti-CRTC3 Rat Monoclonal Antibody [clone: 643729]
Antibodies
Type: Primary
Antigen: CRTC3
Clonality: Monoclonal
Clone: 643729
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR