Anti-SFRP2 Mouse Monoclonal Antibody [clone: 755644]

Supplier: R & D Systems
MAB6838
102886-690EA 594.37 USD
102886-690
Anti-SFRP2 Mouse Monoclonal Antibody [clone: 755644]
Antibodies
Type: Primary
Antigen: SFRP2
Clonality: Monoclonal
Clone: 755644
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR