Anti-PLXNB2 Rat Monoclonal Antibody [clone: 772417]

Supplier: R & D Systems
MAB6836
102886-688EA 594.37 USD
102886-688
Anti-PLXNB2 Rat Monoclonal Antibody [clone: 772417]
Antibodies
Type: Primary
Antigen: PLXNB2
Clonality: Monoclonal
Clone: 772417
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR