Anti-SKP2 Mouse Monoclonal Antibody [clone: 712120]

Supplier: R & D Systems
MAB6817
102886-684EA 577.02 USD
102886-684
Anti-SKP2 Mouse Monoclonal Antibody [clone: 712120]
Antibodies
Type: Primary
Antigen: SKP2
Clonality: Monoclonal
Clone: 712120
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR