Anti-THEMIS Rat Monoclonal Antibody [clone: 719945]

Supplier: R & D Systems
MAB6816
102886-682EA 566.89 USD
102886-682
Anti-THEMIS Rat Monoclonal Antibody [clone: 719945]
Antibodies
Type: Primary
Antigen: THEMIS
Clonality: Monoclonal
Clone: 719945
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR