Anti-AKT1 Mouse Monoclonal Antibody [clone: 7122]

Supplier: R & D Systems
MAB6815
102886-680EA 577.02 USD
102886-680
Anti-AKT1 Mouse Monoclonal Antibody [clone: 7122]
Antibodies
Type: Primary
Antigen: AKT1
Clonality: Monoclonal
Clone: 7122
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR