Anti-CD79B Rat Monoclonal Antibody [clone: 735451]

Supplier: R & D Systems
MAB6814
102886-678EA 594.37 USD
102886-678
Anti-CD79B Rat Monoclonal Antibody [clone: 735451]
Antibodies
Type: Primary
Antigen: CD79B
Clonality: Monoclonal
Clone: 735451
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR