Anti-CCNE1 Mouse Monoclonal Antibody [clone: 713573]

Supplier: R & D Systems
MAB6810
102886-676EA 594.37 USD
102886-676
Anti-CCNE1 Mouse Monoclonal Antibody [clone: 713573]
Antibodies
Type: Primary
Antigen: CCNE1
Clonality: Monoclonal
Clone: 713573
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR