Anti-IL1B1 Mouse Monoclonal Antibody [clone: 77720]

Supplier: R & D Systems
MAB681
220021-424EA 772.25 USD
220021-424
Anti-IL1B1 Mouse Monoclonal Antibody [clone: 77720]
Antibodies
Type: Primary
Antigen: IL1B1
Clonality: Monoclonal
Clone: 77720
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR