Anti-ATG12 Mouse Monoclonal Antibody [clone: 628902]

Supplier: R & D Systems
MAB6807
102886-674EA 594.37 USD
102886-674
Anti-ATG12 Mouse Monoclonal Antibody [clone: 628902]
Antibodies
Type: Primary
Antigen: ATG12
Clonality: Monoclonal
Clone: 628902
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR