Anti-DIAPH2 Mouse Monoclonal Antibody [clone: 699142]

Supplier: R & D Systems
MAB6806
102886-672EA 561.11 USD
102886-672
Anti-DIAPH2 Mouse Monoclonal Antibody [clone: 699142]
Antibodies
Type: Primary
Antigen: DIAPH2
Clonality: Monoclonal
Clone: 699142
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR