Anti-ZNF143 Mouse Monoclonal Antibody [clone: 677314]

Supplier: R & D Systems
MAB6804
102886-670EA 577.02 USD
102886-670
Anti-ZNF143 Mouse Monoclonal Antibody [clone: 677314]
Antibodies
Type: Primary
Antigen: ZNF143
Clonality: Monoclonal
Clone: 677314
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR