Anti-CDKN2B Mouse Monoclonal Antibody [clone: 651308]

Supplier: R & D Systems
MAB6798
102886-666EA 594.37 USD
102886-666
Anti-CDKN2B Mouse Monoclonal Antibody [clone: 651308]
Antibodies
Type: Primary
Antigen: CDKN2B
Clonality: Monoclonal
Clone: 651308
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR