Anti-NLRP3 Mouse Monoclonal Antibody [clone: 652514]

Supplier: R & D Systems
MAB6789
102886-664EA 594.37 USD
102886-664
Anti-NLRP3 Mouse Monoclonal Antibody [clone: 652514]
Antibodies
Type: Primary
Antigen: NLRP3
Clonality: Monoclonal
Clone: 652514
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR