Anti-MEF2C Mouse Monoclonal Antibody [clone: 681824]

Supplier: R & D Systems
MAB6786
102886-414EA 600.15 USD
102886-414
Anti-MEF2C Mouse Monoclonal Antibody [clone: 681824]
Antibodies
Type: Primary
Antigen: MEF2C
Clonality: Monoclonal
Clone: 681824
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR