Anti-FOXA1 Mouse Monoclonal Antibody [clone: 654126]

Supplier: R & D Systems
MAB6778
102886-412EA 594.37 USD
102886-412
Anti-FOXA1 Mouse Monoclonal Antibody [clone: 654126]
Antibodies
Type: Primary
Antigen Symbol: FOXA1
Antigen Name: forkhead box A1
Antigen Synonyms: HNF3A, TCF3A
Clonality: Monoclonal
Clone: 654126
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR