Anti-PIK3R2 Mouse Monoclonal Antibody [clone: 572001]

Supplier: R & D Systems
MAB6777
102886-410EA 577.02 USD
102886-410
Anti-PIK3R2 Mouse Monoclonal Antibody [clone: 572001]
Antibodies
Type: Primary
Antigen: PIK3R2
Clonality: Monoclonal
Clone: 572001
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR