Anti-CLEC9A Rat Monoclonal Antibody [clone: 7H11]

Supplier: R & D Systems
MAB67761
102886-408EA 594.37 USD
102886-408
Anti-CLEC9A Rat Monoclonal Antibody [clone: 7H11]
Antibodies
Type: Primary
Antigen: CLEC9A
Clonality: Monoclonal
Clone: 7H11
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR