Anti-CLEC9A Rat Monoclonal Antibody [clone: 700517]

Supplier: R & D Systems
MAB6776
102886-406EA 594.37 USD
102886-406
Anti-CLEC9A Rat Monoclonal Antibody [clone: 700517]
Antibodies
Type: Primary
Antigen: CLEC9A
Clonality: Monoclonal
Clone: 700517
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR