Anti-IL1RAP Mouse Monoclonal Antibody [clone: 89412]

Supplier: R & D Systems
MAB676
102886-402EA 655.11 USD
102886-402
Anti-IL1RAP Mouse Monoclonal Antibody [clone: 89412]
Antibodies
Type: Primary
Antigen: IL1RAP
Clonality: Monoclonal
Clone: 89412
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR