Anti-GP6 Rat Monoclonal Antibody [clone: 784808]

Supplier: R & D Systems
MAB6758
102886-400EA 594.37 USD
102886-400
Anti-GP6 Rat Monoclonal Antibody [clone: 784808]
Antibodies
Type: Primary
Antigen Symbol: GP6
Antigen Name: glycoprotein 6 (platelet)
Antigen Synonyms: 9830166G18Rik, Gm469, Gpvi
Clonality: Monoclonal
Clone: 784808
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR