Anti-LILRA5 Mouse Monoclonal Antibody [clone: 711827]

Supplier: R & D Systems
MAB67541
102886-398EA 584.25 USD
102886-398
Anti-LILRA5 Mouse Monoclonal Antibody [clone: 711827]
Antibodies
Type: Primary
Antigen: LILRA5
Clonality: Monoclonal
Clone: 711827
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR