Anti-LILRA5 Mouse Monoclonal Antibody [clone: 711828]

Supplier: R & D Systems
MAB6754
102886-396EA 594.37 USD
102886-396
Anti-LILRA5 Mouse Monoclonal Antibody [clone: 711828]
Antibodies
Type: Primary
Antigen: LILRA5
Clonality: Monoclonal
Clone: 711828
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR