Anti-LYAR Mouse Monoclonal Antibody [clone: 735838]

Supplier: R & D Systems
MAB6748
102886-394EA 561.11 USD
102886-394
Anti-LYAR Mouse Monoclonal Antibody [clone: 735838]
Antibodies
Type: Primary
Antigen: LYAR
Clonality: Monoclonal
Clone: 735838
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR